Learn More
AdipoGen Siamycin I
SDP
Catalog No. 502562652
Encompass_Preferred
Chemical. CAS 164802-68-0. Formula C97H131N23O26S4. MW 2163.5. Isolated from Streptomyces sp. Antibiotic. Lariat peptide antiviral. Anti-HIV and anti-HSV Herpes simplex virus agent. Fsr quorum sensing inhibitor. Calmodulin-activated myosin light chain kinase MLCK inhibitor.
Catalog No. 50-256-2652 Supplier AdipoGen Supplier No. AGCN20146M001
Add to Cart
Add to Cart
Chemical. CAS 164802-68-0. Formula C97H131N23O26S4. MW 2163.5. Isolated from Streptomyces sp. Antibiotic. Lariat peptide antiviral. Anti-HIV and anti-HSV Herpes simplex virus agent. Fsr quorum sensing inhibitor. Calmodulin-activated myosin light chain kinase MLCK inhibitor.