Learn More
Apexbio Technology LLC SIB 1553A hydrochloride 191611-89-9 10mg
SDP
Catalog No. 502240266
Encompass_Preferred
SIB 1553A hydrochloride (CAS 191611-89-9) is a selective agonist of nicotinic acetylcholine receptors (nAChRs) containing the 4 subunit This compound exhibits oral bioavailability and modulates central cholinergic neurotransmission by selectively activating 4-containing nAChRs Preclinical studies indicate that SIB 1553A hydrochloride enhances cognitive performance likely through its influence on neuronal signaling pathways in the central nervous system It serves as a valuable tool for investigating the role of specific nAChR subtypes in cognition and is under evaluation for its potential in symptomatic research on Alzheimer s disease and other cognitive disorders
This item is not returnable. View return policy
Catalog No. 50-224-0266 Supplier Apexbio Technology LLC Supplier No. B570810
SIB 1553A hydrochloride (CAS 191611-89-9) is a selective agonist of nicotinic acetylcholine receptors (nAChRs) containing the 4 subunit This compound exhibits oral bioavailability and modulates central cholinergic neurotransmission by selectively activating 4-containing nAChRs Preclinical studies indicate that SIB 1553A hydrochloride enhances cognitive performance likely through its influence on neuronal signaling pathways in the central nervous system It serves as a valuable tool for investigating the role of specific nAChR subtypes in cognition and is under evaluation for its potential in symptomatic research on Alzheimer s disease and other cognitive disorders