Learn More
Medchemexpress LLC HY-18684 5mg Medchemexpress, SIBA CAS:35899-54-8 Purity:>98%
SDP
Catalog No. 502082069
Encompass_Preferred
Medchemexpress, HY-18684 5mg SIBA CAS:35899-54-8 SIBA (5'-Isobutylthioadenosine) is a synthetic analogue of S-adenosylhomocysteine (SAH, HY-19528) and acts as an inhibitor of SAM-mediated transmethylation. SIBA can interfere with a variety of enzymatic activities in vitro, such as S-adenosylhomocysteine hydrolase, methylthioadenosine phosphorylase and cAMP phosphodiesterase. SIBA reversibly blocks the multiplication of herpes simplex type 1 virus (HSV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2069 Supplier Medchemexpress LLC Supplier No. HY186845MG
Medchemexpress, HY-18684 5mg SIBA CAS:35899-54-8 SIBA (5'-Isobutylthioadenosine) is a synthetic analogue of S-adenosylhomocysteine (SAH, HY-19528) and acts as an inhibitor of SAM-mediated transmethylation. SIBA can interfere with a variety of enzymatic activities in vitro, such as S-adenosylhomocysteine hydrolase, methylthioadenosine phosphorylase and cAMP phosphodiesterase. SIBA reversibly blocks the multiplication of herpes simplex type 1 virus (HSV). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.