Learn More
Vee Gee Vee Gee Glass Beaker 100 mL
Catalog No. NC1937411
Encompass_Preferred
The low-form Griffin beaker is made from high-quality borosilicate tubing and features an easy-pour spout and permanent white graduations. Uniform wall thickness - ideal for heating applications Made from high-quality borosilicate tubing with a low coefficient of expansion Graduation Accuracy: 5% of full capacity Graduation Range: 20 to 100 mL Graduation Interval: 10 mL
Catalog No. NC1937411 Supplier Vee Gee Supplier No. 10020100A
The low-form Griffin beaker is made from high-quality borosilicate tubing and features an easy-pour spout and permanent white graduations. Uniform wall thickness - ideal for heating applications Made from high-quality borosilicate tubing with a low coefficient of expansion Graduation Accuracy: 5% of full capacity Graduation Range: 20 to 100 mL Graduation Interval: 10 mL