Learn More
Vee Gee Glass Buret, 50 mL
Catalog No. NC2386481
Encompass_Preferred
Designed """"to deliver"""" the Glass Buret accurately dispenses liquid and meets ASTM E-287 Class A specifications. • Made from high-quality borosilicate tubing with a low coefficient of expansion• Features durable, permanent markings; fine, sharp lines; and large easy-to-read numbers• Supplied with PTFE stopcock to eliminate freezing and lubricant contamination
Catalog No. NC2386481 Supplier Vee Gee Supplier No. 2131050
Designed """"to deliver"""" the Glass Buret accurately dispenses liquid and meets ASTM E-287 Class A specifications. • Made from high-quality borosilicate tubing with a low coefficient of expansion• Features durable, permanent markings; fine, sharp lines; and large easy-to-read numbers• Supplied with PTFE stopcock to eliminate freezing and lubricant contamination