Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sibiricaxanthone B | 241125-81-5 | 5 MG
SDP

Catalog No. 5000419195
Encompass_Preferred

Sibiricaxanthone B is a xanthone isolated from Polygala tenuifolia, intended for research use. Studies have shown that when orally administered after Kai-Xin-San, Sibiricaxanthone B exhibited no significant differences in pharmacokinetic parameters in normal and AD rats.

  • Purity of 99.12%
  • Appears as a solid
  • Color is light yellow to green yellow
  • Isolated from Polygala tenuifolia
  • Suitable for research applications

Catalog No. 50-004-19195 Supplier Medchemexpress LLC Supplier No. HYN21715MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sibiricaxanthone B is a xanthone isolated from Polygala tenuifolia, intended for research use. Studies have shown that when orally administered after Kai-Xin-San, Sibiricaxanthone B exhibited no significant differences in pharmacokinetic parameters in normal and AD rats.

  • Purity of 99.12%
  • Appears as a solid
  • Color is light yellow to green yellow
  • Isolated from Polygala tenuifolia
  • Suitable for research applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.