Learn More
Medchemexpress LLC Sibiricose A5 | 107912-97-0 | 20 MG
SDP
Catalog No. 5000419192
Encompass_Preferred
Sibiricose A5 is an oligosaccharide ester isolated from Polygalae Radix with potent antioxidant activity. It is for research use only and not sold to patients.
  • Isolated from Polygalae Radix.
  • Possesses potent antioxidant activity.
  • High purity (98.67%).
  • Available in various quantities.
Catalog No. 50-004-19192 Supplier Medchemexpress LLC Supplier No. HYN216720MG
Sibiricose A5 is an oligosaccharide ester isolated from Polygalae Radix with potent antioxidant activity. It is for research use only and not sold to patients.
  • Isolated from Polygalae Radix.
  • Possesses potent antioxidant activity.
  • High purity (98.67%).
  • Available in various quantities.