Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sibiricose A5 | 107912-97-0 | 5 MG
SDP

Catalog No. 5000418283
Encompass_Preferred

Sibiricose A5 is an oligosaccharide ester isolated from Polygalae Radix, possessing potent antioxidant activity. It is intended for research use only.

  • Isolated from Polygalae Radix
  • Possesses potent antioxidant activity
  • Appears as an off-white to light yellow solid
  • Molecular formula C22H30O14
  • Molecular weight 518.47
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in DMSO (50 mg/mL)

Catalog No. 50-004-18283 Supplier Medchemexpress LLC Supplier No. HYN21675MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sibiricose A5 is an oligosaccharide ester isolated from Polygalae Radix, possessing potent antioxidant activity. It is intended for research use only.

  • Isolated from Polygalae Radix
  • Possesses potent antioxidant activity
  • Appears as an off-white to light yellow solid
  • Molecular formula C22H30O14
  • Molecular weight 518.47
  • Stable under recommended storage conditions
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in DMSO (50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.