Learn More
Medchemexpress LLC Sibrafiban (RO 48-3657) | 172927-65-0 | 99.7% | 420.47 g/mol | C20H28N4O6 | 5 MG
SDP
Catalog No. 5000247927
Encompass_Preferred
Sibrafiban (RO 48-3657) is an orally active, nonpeptide double prodrug of Ro 44-3888 that functions as a selective glycoprotein IIb/IIIa receptor antagonist. It inhibits platelet aggregation and is supplied as a high-purity research chemical for in vitro and preclinical studies.
  • Orally active double prodrug with selective glycoprotein IIb/IIIa antagonism.
  • Inhibits platelet aggregation for antithrombotic research.
  • High purity (≈99.7%).
  • Provided as a solid research reagent suitable for in vitro and preclinical assays.
  • Recommended powder storage: -20°C for long-term stability.
Catalog No. 50-002-47927 Supplier Medchemexpress LLC Supplier No. HY103095MG
Sibrafiban (RO 48-3657) is an orally active, nonpeptide double prodrug of Ro 44-3888 that functions as a selective glycoprotein IIb/IIIa receptor antagonist. It inhibits platelet aggregation and is supplied as a high-purity research chemical for in vitro and preclinical studies.
  • Orally active double prodrug with selective glycoprotein IIb/IIIa antagonism.
  • Inhibits platelet aggregation for antithrombotic research.
  • High purity (≈99.7%).
  • Provided as a solid research reagent suitable for in vitro and preclinical assays.
  • Recommended powder storage: -20°C for long-term stability.