Learn More
Medchemexpress LLC SIC-19 | 713112-69-7 | 98.0% | 574.67 | 25 MG
SDP
Catalog No. 5000392540
Encompass_Preferred
This product is a SIK2 inhibitor that promotes SIK2 protein degradation through the ubiquitination pathway. It has been shown to inhibit cancer cell growth and sensitize cells to PARP inhibitors, as well as in ovarian cancer organoids and xenograft models. It is intended for research use only.
  • SIK2 inhibitor
  • Promotes SIK2 protein degradation via the ubiquitination pathway
  • Inhibits cancer cell growth
  • Sensitizes cells to PARP inhibitors
  • Effective in ovarian cancer organoids and xenograft models
Catalog No. 50-003-92540 Supplier Medchemexpress LLC Supplier No. 25MGHY163743
This product is a SIK2 inhibitor that promotes SIK2 protein degradation through the ubiquitination pathway. It has been shown to inhibit cancer cell growth and sensitize cells to PARP inhibitors, as well as in ovarian cancer organoids and xenograft models. It is intended for research use only.
  • SIK2 inhibitor
  • Promotes SIK2 protein degradation via the ubiquitination pathway
  • Inhibits cancer cell growth
  • Sensitizes cells to PARP inhibitors
  • Effective in ovarian cancer organoids and xenograft models