Learn More
Medchemexpress LLC SIC-19 | 713112-69-7 | 98.0% | 574.67 | 5 MG
SDP
Catalog No. 5000391191
Encompass_Preferred
SIC-19 is a SIK2 inhibitor that promotes SIK2 protein degradation via the ubiquitination pathway. It inhibits cancer cell growth and sensitizes cells to PARP inhibitors, and has shown effectiveness in ovarian cancer organoids and xenograft models.
  • SIK2 inhibitor
  • Promotes SIK2 protein degradation via the ubiquitination pathway
  • Inhibits cancer cell growth
  • Sensitizes cells to PARP inhibitors
  • Effective in ovarian cancer organoids and xenograft models
Catalog No. 50-003-91191 Supplier Medchemexpress LLC Supplier No. HY1637435MG
SIC-19 is a SIK2 inhibitor that promotes SIK2 protein degradation via the ubiquitination pathway. It inhibits cancer cell growth and sensitizes cells to PARP inhibitors, and has shown effectiveness in ovarian cancer organoids and xenograft models.
  • SIK2 inhibitor
  • Promotes SIK2 protein degradation via the ubiquitination pathway
  • Inhibits cancer cell growth
  • Sensitizes cells to PARP inhibitors
  • Effective in ovarian cancer organoids and xenograft models