Learn More
Medchemexpress LLC Sic-19 | 713112-69-7 | 98.0% | 574.67 g/mol | C29H26N4O5S2 | 10 MG
SDP
Catalog No. 5000289071
Encompass_Preferred
SIC-19 is a small-molecule SIK2 inhibitor that promotes SIK2 protein degradation via the ubiquitination pathway. It inhibits cancer cell growth and sensitizes cells to PARP inhibitors in ovarian cancer organoids and xenograft models. The compound is supplied as a solid, white to light yellow powder with high purity for research applications.
  • Promotes SIK2 protein degradation via the ubiquitination pathway.
  • Inhibits cancer cell growth and sensitizes cells to PARP inhibitors in cellular and in vivo models.
  • High purity: 98.0%.
  • Solid powder, white to light yellow; suitable for biochemical and cellular studies.
  • Stable when stored as powder at -20°C; solution storage at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C29H26N4O5S2; molecular weight 574.67 g/mol.
Catalog No. 50-002-89071 Supplier Medchemexpress LLC Supplier No. HY16374310MG
SIC-19 is a small-molecule SIK2 inhibitor that promotes SIK2 protein degradation via the ubiquitination pathway. It inhibits cancer cell growth and sensitizes cells to PARP inhibitors in ovarian cancer organoids and xenograft models. The compound is supplied as a solid, white to light yellow powder with high purity for research applications.
  • Promotes SIK2 protein degradation via the ubiquitination pathway.
  • Inhibits cancer cell growth and sensitizes cells to PARP inhibitors in cellular and in vivo models.
  • High purity: 98.0%.
  • Solid powder, white to light yellow; suitable for biochemical and cellular studies.
  • Stable when stored as powder at -20°C; solution storage at -80°C (6 months) or -20°C (1 month).
  • Molecular formula C29H26N4O5S2; molecular weight 574.67 g/mol.