Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Sickle Cell Gene Detection (DNA-based) with SYBR™ Safe
SDPSCI_ED

Catalog No. 01672280 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
01-672-280 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-280 Supplier EDVOTEK™ Supplier No. 116S
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this Sickle Cell Gene Detection (DNA-based) with SYBR™ Safe kit, your students investigate the restriction enzyme that discriminates between HbA (normal) and HbS (disease) genes.

  • Includes SYBR™ Safe DNA Stain, NOT FlashBlue™ DNA Stain
  • Sickle Cell Anemia is a common genetic disease that causes long rods in red blood cells, giving them a 'sickled' appearance.
  • Discover how a single nucleotide mutation can profoundly affect a person's health
  • Explore Mendelian genetics and the molecule methods used to identify sickle cell disease
  • Perform agarose gel electrophoresis to separate differently-sized DNA molecules
  • Load, run, analyze, and size control samples and samples from a family of three to identify mutations in their hemoglobin genes
  • Group Size: For 8 Gels
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe DNA Stain, Practice Gel Loading Solution.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade 8 to Undergraduate
Class Size 8 Lab Groups
Product Type Sickle-cell Gene Detection (DNA-based) Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.