Learn More
SURE TECH DIAGNOSTIC ASSOC INC ST 301-A Sickle Cell Test Kit (25 tests)
Catalog No. NC2464901
Encompass_Preferred
Sickle Cell Test Kits. Simple turbidity test for detection of hemoglobin S in anticoagulated blood. 25 test kit
Catalog No. NC2464901 Supplier SURE TECH DIAGNOSTIC ASSOC INC Supplier No. ST 301A
Add to Cart
Add to Cart
Sickle Cell Test Kits. Simple turbidity test for detection of hemoglobin S in anticoagulated blood. 25 test kit