Learn More
Great Lakes Diagnostics SickleHeme™ Sickle Cell Control Kits
GSA_VA
Catalog No. MHK118A
Change view
Click to view available options
Quantity:
60 Tests
Catalog No. Quantity
MH-K118A 60 Tests
1 options
This item is not returnable. View return policy
Catalog No. MH-K118A Supplier Great Lakes Diagnostics Supplier No. GLD1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
two vials each of freezed-dried RBC of Positive(A/S) and Negative(A/A); two vials of diluent; 60 determinations in each kit
Control for the SickleHeme Sickle Cell Solubility Test Kit
  • Shelf life of the control is three years and the reconstituted control is 60 days
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Bi-level Control Kit
Includes 2 Vials each of freezed-dried RBC of positive(A/S) and negative(A/A), 2 vials of diluent
Quantity 60 Tests