Learn More
Great Lakes Diagnostics SickleHeme™ Sickle Cell Solubility Test Kits
Catalog No. MHK120A
Change view
Click to view available options
Quantity:
100 Tests
250 Tests
Includes:
1 Buffer, powder reagent kit
2 Sets of Buffer and powder
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Includes
MH-K120A 250 Tests 1 Buffer, powder reagent kit
MH-K110A 100 Tests 2 Sets of Buffer and powder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MH-K120A Supplier Great Lakes Diagnostics Supplier No. GLD250
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
two sets of buffer and powder, which allows for 90 days coverage from one kit (12- and 100-test); one bottle buffer and powder reagent kit for high volume users (250-test)
All unreconstituted sickle cell solubility kits have a shelf life of two years at room temperature
Specifications
Type Sickle Cell Solubility Test Kit
Includes 1 Buffer, powder reagent kit
Quantity 250 Tests