Learn More
Medchemexpress LLC HY-103353 1mg Medchemexpress, SID 26681509 CAS:958772-66-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1033531MG
Encompass_Preferred
Medchemexpress, HY-103353 1mg SID 26681509 CAS:958772-66-2 SID 26681509 is a potent, reversible, competitive, and selective inhibitor of human cathepsin L with an IC50 of 56 nM. SID 26681509 inhibits in vitro propagation of malaria parasite Plasmodium falciparum and inhibits Leishmania major with IC50s of 15.4 μM and 12.5 μM, respectively. SID 26681509 shows no inhibitory activity against cathepsin G. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8080