Learn More
TARGETMOL CHEMICALS INC SID 26681509 5MG
SDP
Catalog No. 502581403
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SID 26681509 is a selective reversible and competitive human cathepsin L inhibitor (IC50 of 56 nM). purity: 98%
Catalog No. 50-258-1403 Supplier TARGETMOL CHEMICALS INC Supplier No. T129095MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SID 26681509 is a selective reversible and competitive human cathepsin L inhibitor (IC50 of 56 nM). purity: 98%