Learn More
Medchemexpress LLC Sid 26681509 quarterhydrate | 00-00-0 | 98.0% | 544.16 g/mol | C27H33N5O5S.1/4H2O | 5 MG
SDP
Catalog No. 5000244872
Encompass_Preferred
Potent, reversible, competitive inhibitor of human cathepsin L (IC50 = 56 nM) with reported antiparasitic activity in vitro. Supplied as a white to off-white solid with high reported purity, the compound is provided in milligram quantities and as a ready-to-use DMSO solution for biochemical and antiparasitic research.
  • Selective cathepsin L inhibition (IC50 = 56 nM).
  • Inhibits Plasmodium falciparum and Leishmania major in vitro.
  • High reported purity (98.02%).
  • White to off-white solid; also available as a 10 mM solution in DMSO.
  • Available in small milligram package sizes suitable for biochemical assays.
Catalog No. 50-002-44872 Supplier Medchemexpress LLC Supplier No. HY103353A5MG
Potent, reversible, competitive inhibitor of human cathepsin L (IC50 = 56 nM) with reported antiparasitic activity in vitro. Supplied as a white to off-white solid with high reported purity, the compound is provided in milligram quantities and as a ready-to-use DMSO solution for biochemical and antiparasitic research.
  • Selective cathepsin L inhibition (IC50 = 56 nM).
  • Inhibits Plasmodium falciparum and Leishmania major in vitro.
  • High reported purity (98.02%).
  • White to off-white solid; also available as a 10 mM solution in DMSO.
  • Available in small milligram package sizes suitable for biochemical assays.