Learn More
Cayman Chemical SID 3712249, Purity: ≥95%, CAS: 522606-67-3, 50 mg
SDP
Catalog No. 502988783
Encompass_Preferred
An inhibitor of miR-544 biogenesis; increases MTOR mRNA expression and decreases the mRNA expression of the genes encoding miR-544, HIF-1α, and ATM in MDA-MB-231, MCF-7, and MCF-10A cells under hypoxic conditions at 20 nM; induces apoptosis in MDA-MB-231 cells in a hypoxia- and mTOR-dependent manner at 20 nM; enhances the senstivity of MCF-7 and MDA-MB-231 cells cultured under hypoxic conditions to 5-FU-induced decreases in cell viability at 20 nM; reduces tumor growth in an MDA-MB-231 mouse xenograft model
Catalog No. 50-298-8783 Supplier Cayman Chemical Supplier No. 2972350 MG
An inhibitor of miR-544 biogenesis; increases MTOR mRNA expression and decreases the mRNA expression of the genes encoding miR-544, HIF-1α, and ATM in MDA-MB-231, MCF-7, and MCF-10A cells under hypoxic conditions at 20 nM; induces apoptosis in MDA-MB-231 cells in a hypoxia- and mTOR-dependent manner at 20 nM; enhances the senstivity of MCF-7 and MDA-MB-231 cells cultured under hypoxic conditions to 5-FU-induced decreases in cell viability at 20 nM; reduces tumor growth in an MDA-MB-231 mouse xenograft model