Learn More
Medchemexpress LLC SID7970631 | 868224-75-3 | 98.3% | 100 MG
SDP
Catalog No. 5000340626
Encompass_Preferred
SID7970631 is an isoquinolinone analog and a potent, selective submicromolar SF-1 inhibitor with an IC50 of 255 nM. It is primarily used for cancer research and exhibits various cytotoxic effects in vitro.
  • Potent and selective SF-1 inhibitor (IC50 = 255 nM)
  • Isoquinolinone analog
  • Useful in cancer research
  • Exhibits half-maximal cytotoxic effect in CHO-K1 cells
  • Dose-dependently inhibits luciferase expression in SF-1 assay
  • Demonstrates cytotoxic effect on H295R/TR SF-1 cells and SW-13 cells
Catalog No. 50-003-40626 Supplier Medchemexpress LLC Supplier No. HY128207100MG
SID7970631 is an isoquinolinone analog and a potent, selective submicromolar SF-1 inhibitor with an IC50 of 255 nM. It is primarily used for cancer research and exhibits various cytotoxic effects in vitro.
  • Potent and selective SF-1 inhibitor (IC50 = 255 nM)
  • Isoquinolinone analog
  • Useful in cancer research
  • Exhibits half-maximal cytotoxic effect in CHO-K1 cells
  • Dose-dependently inhibits luciferase expression in SF-1 assay
  • Demonstrates cytotoxic effect on H295R/TR SF-1 cells and SW-13 cells