Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SID7970631 | 868224-75-3 | 98.3% | 50 MG
SDP

Catalog No. 5000339348
Encompass_Preferred

SID7970631, an isoquinolinone analog, is a potent and selective submicromolar SF-1 inhibitor with an IC50 of 255 nM. It is used for cancer research.

  • Potent and selective submicromolar SF-1 inhibitor (IC50 of 255 nM)
  • Used for cancer research
  • Exhibits a half-maximal cytotoxic effect in CHO-K1 cells
  • Dose-dependently inhibits luciferase expression in the SF-1 assay
  • Shows cytotoxic effects on H295R/TR SF-1 cells and SW-13 cells
  • Molecular weight is 452.46 with the formula C24H24N2O7
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-39348 Supplier Medchemexpress LLC Supplier No. HY12820750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SID7970631, an isoquinolinone analog, is a potent and selective submicromolar SF-1 inhibitor with an IC50 of 255 nM. It is used for cancer research.

  • Potent and selective submicromolar SF-1 inhibitor (IC50 of 255 nM)
  • Used for cancer research
  • Exhibits a half-maximal cytotoxic effect in CHO-K1 cells
  • Dose-dependently inhibits luciferase expression in the SF-1 assay
  • Shows cytotoxic effects on H295R/TR SF-1 cells and SW-13 cells
  • Molecular weight is 452.46 with the formula C24H24N2O7
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.