Learn More
Medchemexpress LLC SID7970631 | 868224-75-3 | 98.30% | 10 MG
SDP
Catalog No. 5000341244
Encompass_Preferred
SID7970631 is an isoquinolinone analog that functions as a potent and selective submicromolar SF-1 inhibitor (IC50=255 nM). It can be used for cancer research.
  • Potent and selective submicromolar SF-1 inhibitor
  • IC50 of 255 nM (SF-1)
  • Displays cytotoxic effects on H295R/TR SF-1 cells and SW-13 cells
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-41244 Supplier Medchemexpress LLC Supplier No. HY12820710MG
SID7970631 is an isoquinolinone analog that functions as a potent and selective submicromolar SF-1 inhibitor (IC50=255 nM). It can be used for cancer research.
  • Potent and selective submicromolar SF-1 inhibitor
  • IC50 of 255 nM (SF-1)
  • Displays cytotoxic effects on H295R/TR SF-1 cells and SW-13 cells
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month