Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sid7970631 | 868224-75-3 | MFCD04473864 | 98.3% | 452.46 g/mol | C24H24N2O7 | 5 MG
SDP

Catalog No. 5000264828
Encompass_Preferred

SID7970631 is an isoquinolinone analog reported as a potent and selective submicromolar inhibitor of Steroidogenic Factor-1 (SF-1) with an IC50 of 255 nM. Supplied as a solid research compound, it is intended for use in studies of SF-1 signaling and cancer-related cellular assays.

  • Potent SF-1 inhibitor (IC50 255 nM).
  • Submicromolar activity suitable for cell-based assays.
  • Solid powder with high purity (98.3%).
  • Molecular formula C24H24N2O7; molecular weight 452.46 g/mol.
  • Storage recommendations: powder -20°C long term, 4°C short term; solutions -80°C for months.
  • Available as small-scale solids and 10 mM solution in DMSO for research use.

Catalog No. 50-002-64828 Supplier Medchemexpress LLC Supplier No. HY1282075MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SID7970631 is an isoquinolinone analog reported as a potent and selective submicromolar inhibitor of Steroidogenic Factor-1 (SF-1) with an IC50 of 255 nM. Supplied as a solid research compound, it is intended for use in studies of SF-1 signaling and cancer-related cellular assays.

  • Potent SF-1 inhibitor (IC50 255 nM).
  • Submicromolar activity suitable for cell-based assays.
  • Solid powder with high purity (98.3%).
  • Molecular formula C24H24N2O7; molecular weight 452.46 g/mol.
  • Storage recommendations: powder -20°C long term, 4°C short term; solutions -80°C for months.
  • Available as small-scale solids and 10 mM solution in DMSO for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.