Learn More
Ace Glass, Inc. Side arm adapter, 75°, 24/40 inner joints bottom and side, 24/40 outer joint top
SDP
Catalog No. NC0253117
Encompass_Preferred
Side arm adapter, 75°, 24/40 inner joints bottom and side, 24/40 outer joint top
Catalog No. NC0253117 Supplier Ace Glass, Inc. Supplier No. 504010
Side arm adapter, 75°, 24/40 inner joints bottom and side, 24/40 outer joint top