Learn More
Caron Products And Services Side Mounted Carboy for Humidified Chambers
Catalog No. NC2462898
Encompass_Preferred
Side Mounted Carboy for Humidified Chambers. Water fill bottle with cap, 2.5 gallons (9.5 liters). Includes tubing and fittings.
Catalog No. NC2462898 Supplier Caron Products And Services Supplier No. BOTL301
Side Mounted Carboy for Humidified Chambers. Water fill bottle with cap, 2.5 gallons (9.5 liters). Includes tubing and fittings.