Learn More
Grainger Side Outlet Fr Pipe Sz 2 in
Catalog No. 502749182
Encompass_Preferred
The Grainger 14G939 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and delay, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery assist to ensure the auto-darkening filter works even in low light conditions. The Speedglas helmet is designed for reliability and versatility on the job.
Catalog No. 50-274-9182 Supplier Grainger Supplier No. 14G939
The Grainger 14G939 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and delay, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery assist to ensure the auto-darkening filter works even in low light conditions. The Speedglas helmet is designed for reliability and versatility on the job.