Learn More
Chemglass Life Sciences Side Support Shelf, PTFE Coated Aluminum, Non-Folding, 12-7/8" W x 13" L
SDP
Catalog No. 501531342
Encompass_Preferred
Side Support Shelf, PTFE Coated Aluminum, Non-Folding, 12-7/8" W x 13" L. Removable side shelves for Chemglass reactor support frames offer convenient support for the Chemglass data logger, motor controller, temperature monitor/controller, reagent bottles, etc. Shelves attach to the uprights of the support frame via PTFE coated aluminum mounting brackets. Shelves are constructed of aluminum and PTFE coated for chemical resistance. Units can be mounted on either side of the reactor frame. PLEASE NOTE: THE CG-1968-X-50 IS DESIGNED TO SUPPORT THE CG-1972-65 TEMPERATURE CONTROLLER.
Catalog No. 50-153-1342 Supplier Chemglass Life Sciences Supplier No. CG1968X50
Side Support Shelf, PTFE Coated Aluminum, Non-Folding, 12-7/8" W x 13" L. Removable side shelves for Chemglass reactor support frames offer convenient support for the Chemglass data logger, motor controller, temperature monitor/controller, reagent bottles, etc. Shelves attach to the uprights of the support frame via PTFE coated aluminum mounting brackets. Shelves are constructed of aluminum and PTFE coated for chemical resistance. Units can be mounted on either side of the reactor frame. PLEASE NOTE: THE CG-1968-X-50 IS DESIGNED TO SUPPORT THE CG-1972-65 TEMPERATURE CONTROLLER.