Learn More
Grainger Condor Sideshield, Clear Plastic, Slip-On, ANSI Z87.1, 1 Pair
Catalog No. NC0579634
Encompass_Preferred
These clear plastic sideshields provide additional protection for safety eyewear and prescription spectacles. Designed for versatile use, these slip-on shields are compatible with universal fit eyewear styles to enhance overall eye safety.
  • Slip-on attachment for easy installation
  • Constructed from clear plastic material
  • Features no metal parts for safety in sensitive environments
  • Meets ANSI Z87.1 standards
  • Dimensions are 1 1/2 in. wide by 7/8 in. high by 1 3/4 in. deep
Catalog No. NC0579634 Supplier Grainger Supplier No. 3BE76
These clear plastic sideshields provide additional protection for safety eyewear and prescription spectacles. Designed for versatile use, these slip-on shields are compatible with universal fit eyewear styles to enhance overall eye safety.
  • Slip-on attachment for easy installation
  • Constructed from clear plastic material
  • Features no metal parts for safety in sensitive environments
  • Meets ANSI Z87.1 standards
  • Dimensions are 1 1/2 in. wide by 7/8 in. high by 1 3/4 in. deep