Learn More
Agilent Technologies SIDE -STEP
Catalog No. NC9332800
Encompass
SideStep Lysis and Stabilization Buffer, 100 rxns, Cell lysis and immediate stabilization of RNA and DNA for real time PCR analysis., 10mL
Catalog No. NC9332800 Supplier Agilent Technologies Supplier No. 400900
SideStep Lysis and Stabilization Buffer, 100 rxns, Cell lysis and immediate stabilization of RNA and DNA for real time PCR analysis., 10mL