Learn More
Chemglass Life Sciences Side Swivel Support for Diagonal Distillation on CG-1968-X Support Stand
SDP
Catalog No. 5000980591
Encompass_Preferred
Diagonal distillation cart system for Chemglass reactor systems. Supplied complete with glassware, swivel support assembly and cart system to fit CG-1968-X style frames. The components highlighted in the link below. This add-on kit makes it easy to convert any of our standard reactors over to a complete distillation system.30/50L diagonal distillation cart systems shown in photos. Please Note: Reactor System Must Be Purchased Separately.
Catalog No. 50-009-80591 Supplier Chemglass Life Sciences Supplier No. CG1968D125
Diagonal distillation cart system for Chemglass reactor systems. Supplied complete with glassware, swivel support assembly and cart system to fit CG-1968-X style frames. The components highlighted in the link below. This add-on kit makes it easy to convert any of our standard reactors over to a complete distillation system.30/50L diagonal distillation cart systems shown in photos. Please Note: Reactor System Must Be Purchased Separately.