Learn More
Electron Microscopy Sciences Disposible Reagent Sidecar Reservoir, 100 ml, Sterile
SDP
Supplier:  Electron Microscopy Sciences 7049540
Encompass_Preferred
Sidecar Reservoir features Two-in-One design for precise reagent adding/mixing. Ideal and efficient to use with small volume control and large volume test reagents. "Sidecar" holds as little as 10 ml of liquid or as much as 20 ml.
Capacity: 100 ml
Sterile: Sterile
Catalog No. 50-334-00