Learn More
Sigma Aldrich Fine Chemicals Biosciences sidEntryTM glove box a cle1EA
Catalog No. 502908842
Encompass
The Scienceware sidEntry glove box is a clear acrylic glove box which features a large side entry gasketed door and a shelf rack to store small items. Glove box stability is assured by four adjustable levelers that also level the box and all its contents. The sidEntry glove box provides an isolated work environment for processing samples or handling air-sensitive materials while maintaining an anaerobic or other gas specific environment within the glove box.8 in. glove ports accept standard dry-box gloves without clamps or O-ringsSmall enough to move from area to areaSide door 17 1/2 in. x 17 1/2 in.
Catalog No. 50-290-8842 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2605171EA
The Scienceware sidEntry glove box is a clear acrylic glove box which features a large side entry gasketed door and a shelf rack to store small items. Glove box stability is assured by four adjustable levelers that also level the box and all its contents. The sidEntry glove box provides an isolated work environment for processing samples or handling air-sensitive materials while maintaining an anaerobic or other gas specific environment within the glove box.8 in. glove ports accept standard dry-box gloves without clamps or O-ringsSmall enough to move from area to areaSide door 17 1/2 in. x 17 1/2 in.