Learn More
Research Products International Corp Sidewall Tough Tages for Laser, Yellow, Fits Micro Well Plates, 0.25 x 1.5, 3120 per Package
SDP
Supplier:  Research Products International Corp 247116Y
Encompass
Sidewall Tough-Tags are slender pre-cut labels designed for micro-well plates, petri dishes, and other containers. Labels withstand incubation, boiling water baths, solvents and temperatures down to -20°C(Roll). Labels adhere to all types of plastic and glass surfaces. White labels accept marking from any writing instrument. Ideal size (0.25" x 1.50") to fit perfectly on the sidewall of a standard micro-well plate, petri dish and more. Allows visual identification of individual plates.
Catalog No. 50-197-8066