Learn More
Bachem Xanthenyl linker resin (200-400 mesh, 0.4-0.7 mmol/g)
SDP
Catalog No. 501947374
Encompass_Preferred
Xanthenyl linker resin (200-400 mesh, 0.4-0.7 mmol/g), (4-(9-Fmoc-amino-xanthen-3-yloxy)butyryl)-4-methyl-benzhydrylamide resin, 1g. Sieber Amide Resin. Sieber amide resin has been developed for the Fmoc-SPPS of fully t-butyl protected peptide amides. Cleavage can be achieved with 1% TFA in methylene chloride. Scavengers may be required. As N-methylation increases the acid-lability of the peptide bond, Malakoutikhah et al. employed Sieber resin for obtaining short peptide amides containing multiple MePhe residues.
Catalog No. 50-194-7374 Supplier Bachem Supplier No. 4026167.0001
Xanthenyl linker resin (200-400 mesh, 0.4-0.7 mmol/g), (4-(9-Fmoc-amino-xanthen-3-yloxy)butyryl)-4-methyl-benzhydrylamide resin, 1g. Sieber Amide Resin. Sieber amide resin has been developed for the Fmoc-SPPS of fully t-butyl protected peptide amides. Cleavage can be achieved with 1% TFA in methylene chloride. Scavengers may be required. As N-methylation increases the acid-lability of the peptide bond, Malakoutikhah et al. employed Sieber resin for obtaining short peptide amides containing multiple MePhe residues.