Learn More
Siemens Healthineers Clinitek Status™+ Analyzer Accessories
GSA_VA
Catalog No. 23042342 Shop All Siemens Healthineers Products
Change view
Click to view available options
Product Type:
Upgrade Kit
Catalog No. Product Type
23-042-342 Upgrade Kit
1 options
Catalog No. 23-042-342 Supplier Siemens Healthineers Supplier No. 1782
Add to Cart
Add to Cart
Upgrade the Status+ Analyzer to the Status Connect System with these accessories.
  • Upgrade Kit includes: 1 connector, 1 Barcode Reader, V2.2 Software
  • Connector accessory is a platform base to which the analyzer can attach to provide the unit with additional connectivity and software features
  • This item is non-returnable.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Upgrade Kit
For Use With (Equipment) Clinitek Status+ Analyzer
Includes 1 Connector, 1 Barcode Reader, V2.2 Software
Quantity Each
WARNING: Reproductive Harm - www.P65Warnings.ca.gov