Learn More
Siemens Healthineers Chek-Stix™ Urinalysis Control Strips
GSA_VA
Catalog No. AM1360 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
25 Pack
Catalog No. Quantity
AM-1360 25 Pack
1 options
Catalog No. AM-1360 Supplier Siemens Healthineers Supplier No. 10310482
Add to Cart
Edge
Add to Cart
Positive results only
  • Firm plastic strips with six distinct analyte areas used to check urinalysis results over time
  • Each area contains one or more natural or synthetic ingredients
  • Control strips are reconstituted with distilled water to provide positive results with each reagent pad on Multistix™ SG Reagent Strips
  • Stable for eight hours after reconstitution
Order Info
Packaged 25 strips/bottle.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
For Use With (Application) Urinalysis
Quantity 25 Pack