Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Clinitek™ Advantus™ Semi-Automated Urine Chemistry Analyzer

Catalog No. 09529370 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-529-370 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-529-370 Supplier Siemens Healthineers Supplier No. 10636672
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Streamlined operation with immediate start-up

  • Easy-to-use color touch screen
  • Ability to consolidate microscopy results onto one patient record
  • Increased control with automatic QC prompting and QC lockout capability
  • New Auto-Checks feature identifies compromised strips by humidity and automatically adjusts
  • Confirmatory and microscopic sieve functions flag abnormal results for quick review
  • Convenient data retrieval for up to 500 patients

  • Automatic calibration improves accuracy for every strip
  • Boosts productivity with throughput of 7 seconds per sample
  • Simple bar code identification of sample, color, clarity, lot number and expiration date values minimizes errors
  • Test format is dry chemistry reagent strips
  • Dual readings at reactive and reference wavelengths reduce urine pigment interference
  • Automatically determines urine color
  • Sample loading is self-paced
  • Sample handling has one-touch STAT interrupt capability
  • Single or batch mode sample processing
  • Sample ID entry can be made manually, via bar code, or downloaded from LIS
  • Network ready

Automatic reading of Multistix™ urinalysis tests, including: Leukocyte, Nitrite, Protein, Blood, Glucose, Ketone, Bilirubin, Urobilinogen, pH, Specific Gravity, Creatinine and Protein-to-Creatinine Ratio

Compliance

UL, CE, EMC

TRUSTED_SUSTAINABILITY

Specifications

Product Type Semi-automated Urine Chemistry Analyzer
Calibration Automatic with every strip
Humidity 20 to 80%
Memory 500 patient records plus 200 controls
Temperature (English) Operating 64°F to 86°F
Temperature (Metric) Operating 18°C to 30°C
Depth (English) 13.75 in.
Depth (Metric) 35 cm
Height (English) 12.75 in.
Height (Metric) 32 cm
Width (English) 15.75 in.
Width (Metric) 39 cm
Weight (English) 16 lb.
Weight (Metric) 7.2 kg
For Use With (Application) Automatic reading of leukocyte, Nitrite, Protein, Blood, Glucose, Ketone, Bilirubin, Urobilinogen, pH, specific gravity, Creatinine and Protein-to-creatinine ratio
Certifications/Compliance UL, CE, EMC
Quantity Each
Electrical Requirements 100/240 V 50/60 Hz
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.