Learn More
Siemens Healthineers Clinitek™ Microalbumin 2 Reagent Strips
GSA_VA
Catalog No. am2085 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
AM-2085 Each
1 options
Catalog No. AM-2085 Supplier Siemens Healthineers Supplier No. 10317439
Add to Cart
Add to Cart
For use with Clinitek 50, 100, Status Family of Urine Chemistry Analyzers
  • Semiquantitative test
  • Detects human albumin and creatinine in urine CLIA waived on the Clinitek 50
  • Provides recommended albumin-to-creatinine results in one minute
  • CLIA waived on Clinitek Status family of analyzers
  • Results from instrument-read strips are printed, eliminating transcription errors
Order Info
Packaged 25 strips/bottle
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Waived
Type Microalbumin Reagent Strip
For Use With (Application) Clinitek™ 50, 100, status+ and status connect Urine chemistry analyzer
Quantity Each