Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Clinitek Status™+ Analyzer

Catalog No. 23042346 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23042346 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23042346 Supplier Siemens Healthineers Supplier No. 10379675
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enjoy quick, accurate and reliable urinalysis results from this easy-to-use, automated analyzer.

The new CLINITEK Status+ Analyzer is the latest addition to the CLINITEK family, the most widely used urinalysis brand worldwide. The Status+ Analyzer is even easier to use and features a new Auto-Checks feature.
The analyzer automatically checks each test strip for humidity exposure and strip identification for Siemens test strips. Together, these provide improved reagent integrity and result accuracy.

Easy to Use and Convenient
  • Simple, intuitive touchscreen operation
  • Load test — the analyzer does the rest, automatically times and reads each test
  • Fast results — in-office test results in about 1 minute
  • Automatic printed report — no manual transcription needed
Improved Data Integrity
  • Analyzer along with patented test strip technology enables Auto-Checks:
  • Checks each strip for humidity to prevent reporting of falsely elevated results
  • Auto-identification of test strips, no need for operators to key in manually
  • Removes subjectivity of visually read results
Comprehensive Menu
  • Routine urinalysis with CLINITEK Multistix™ urine test strips, the most widely used test strips in the world
  • Albumin-to-creatinine ratio test to detect early kidney disease in diabetic and hypertensive patients
  • Protein-to-creatinine ratio test to detect early kidney disease in patients at risk for chronic kidney disease
  • hCG pregnancy testing via the CLINITEST™ hCG cassette
Future Ready
  • Simple connectivity upgrade to CLINITEK Status Connect System, for remote oversight, data integration and elimination of manual documentation when interfaced to data management software, EMR or LIS
  • Barcode reading capability to eliminate manual entry of patient and operator ID
TRUSTED_SUSTAINABILITY

Specifications

Product Type Analyzer
Calibration Automatic, self-calibrating
Humidity 18 to 80%
Memory 950 results
Temperature (English) Operating 64°F to 86°F
Temperature (Metric) Operating 18°C to 30°C
Depth (English) 10.7 in.
Depth (Metric) 27.2 cm
Height (English) 6.2 in.
Height (Metric) 15.8 cm
Width (English) 6.7 in.
Width (Metric) 17.1 cm
Weight (English) 3.65 lb.
Weight (Metric) 1.66 kg
For Use With (Application) Routine Urine tests, kidney disease test, pregnancy test
Quantity Each
Electrical Requirements 100/240 V 50/60 Hz
Frequency 50/60 Hz
Voltage 100/240 V
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.