Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Clinitest™ hCG Test Cassettes

Catalog No. 22247312 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-247-312 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-247-312 Supplier Siemens Healthineers Supplier No. 10310618
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate need for timing results

  • For use on Clinitek™ Status™ Family of Urine Analyzers
TRUSTED_SUSTAINABILITY

Specifications

Accuracy <99%
Content And Storage Room Temperature
Clia Complexity Waived
Detectable Analytes hCG
Sample Type Urine
Sensitivity 25 mIU/mL
Type hCG Test Cassette
Quantity 25 Tests
Sample Size 200 μL
Test Time ≤5 min.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.