Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers DCA Vantage™ Analyzer
GSA_VA

Catalog No. 23312998 Shop All Siemens Healthineers Products
Change view
Click to view available options
Sample Type:
Whole Blood, Urine
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
23-312-998 Whole Blood, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-312-998 Supplier Siemens Healthineers Supplier No. 10282969
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provides precise and accurate quantitative test results for HbA1c in whole blood during the patient visit to help improve decision making

  • Easy-to-use analyzer improves productivity by providing test results during patient visit in 6 to 7 minutes, reducing time needed for patient follow-up
  • No sample or reagent preparation: sample cartridges require no reagent pipetting
  • Analyzer automatically generates test results and provides a printed record that can be affixed to the patient file and/or uploaded to a network or LIS/HIS
  • Color touch screen with 1/4 VGA resolution
  • 4000 patient and/or control records
  • User-definable reference ranges available for HbA1c
  • 1μL of whole blood needed for HbA1c testing
  • Trending graphs enable more comprehensive patient disease management
  • Reagents and control kits are available separately

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Sample Type Whole Blood, Urine
For Use With (Application) Diabetes management
Analytical Method Monoclonal antibody agglutination reaction, Benedict Behre chemical reaction
Certifications/Compliance IFCC
Depth (English) 10.5 in.
Depth (Metric) 27.7 cm
Height (English) 9 in.
Height (Metric) 25.4 cm
Width (English) 11.5 in.
Width (Metric) 28.7 cm
Electrical Requirements 100/240 VAC 50/60 Hz
Frequency 50/60 Hz
Voltage 100/240 VAC
Weight (English) 9 lb.
Weight (Metric) 3.88 kg
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.