Learn More
Siemens Healthineers Multistix PRO™ Reagent Strips
Catalog No. AM1554 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
100 Pack
25 Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM-1554 100 Pack
AM-1566 25 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM-1554 Supplier Siemens Healthineers Supplier No. 10331147
Add to Cart
Edge
Add to Cart
Proprietary dip-and-read tests, Protein Low (Albumin), Protein High, and Creatinine
  • CLIA Complexity: Waived
  • Results in 60 seconds
  • Provide protein-to-creatinine ratio
  • Compatible with Clinitek™ analyzers
  • Improved protein sensitivity (8–15mg/dL)
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Waived
For Use With (Application) Urinalysis
Quantity 100 Pack
Test Time 60 sec.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov