Learn More
Siemens Healthineers Urinalysis Reagent Test Strips
GSA_VA
Catalog No. AM2190 Shop All Siemens Healthineers Products
Change view
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, pH, Protein, Urobilinogen
Blood
Blood Urea, Nitrogen
Blood, Glucose, Ketone, pH, Protein
Blood, Glucose, pH, Protein
Glucose, Protein
Protein
Quantity:
100 Pack
25 Pack
50 Pack
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Quantity
AM-2190 Blood 50 Pack
AM-2830 Blood Urea, Nitrogen 25 Pack
AM-2179 Bilirubin, Blood, Glucose, Ketone, pH, Protein, Urobilinogen 100 Pack
23-042320 Blood, Glucose, Ketone, pH, Protein 100 Pack
23-042321 Blood, Glucose, pH, Protein 100 Pack
AM-2191 Protein 100 Pack
AM-2184 Glucose, Protein 100 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. AM-2190 Supplier Siemens Healthineers Supplier No. 10312568
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Single-use, plastic strips with multiple reagent test areas
  • Ready to use and entirely disposable
  • Dip and read visually for results
  • Some strips can be read instrumentally on Clinitek™ 50, 100, 200, 200+, 500, Advantus, and Status Family of Urine Chemistry analyzers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Blood
Disposable Single-use
For Use With (Application) Urinalysis, in-vitro diagnostic use
Quantity 50 Pack