Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers URIN-TEK™Urine Collection System
GSA_VA

Catalog No. 23375535 Shop All Siemens Healthineers Products
Change view
Click to view available options
Volume (Metric):
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-375535 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-375535 Supplier Siemens Healthineers Supplier No. 10310944
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A disposal system for urine specimen collection, transportation and testing

  • Specimens are collected and analyzed in the same container
  • Tubes are graduated in milliliters and can be centrifuged for microscopic analysis
  • System Includes:
    • Disposable 15mL, graduated plastic tube with plastic snap-cap
    • Self-adhesive identification label
    • Flat-bottom 3-oz. paper collection cup
    • 10-tube disposable tube holder

Order Info

  • Part No. 23-375535: Packaged as five sets of 100 each of the following: tubes, plastic caps, adhesive labels, collection cups and disposable tube holders
  • Part No. 22-375543: Tubes only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Urine Collection System
For Use With (Application) For Urine specimen collection, transportation and testing
Includes Tubes, Plastic caps, adhesive labels, collection cups and disposable tube holders
Volume (Metric) 15 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.