Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Sierra™ Pleated Mask

Catalog No. 19088881 Shop All DuPont Personal Protection Products
Change view
Click to view available options
Sierra™ Pleated Mask
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-088-881 Sierra™ Pleated Mask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-088-881 Supplier DuPont™ Supplier No. ML735WBU000300BB

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sierra™ masks for cleanroom environments provide a balance of bacteria and particle filtration efficiency and breathability.

  • Available in 7 in. and 9 in. sizes
  • Available in blue and white colors
  • Pleated mask design with metal nose piece
  • Available with tie fasteners or ear loops
  • Cone mask option with adjustable nose piece and over the head band
  • Sterile options are available
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Face Mask
Color Blue
Size Standard
Sterility Non-sterile
Disposable Single-use
For Use With (Application) Controlled Environments
Model ML735WBU000300BB
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.