Learn More
Cole-Parmer Cole-Parmer Testing Sieve, SS Frame/SS Wire, 12" OD, Full height, No. 10
Catalog No. NC1476301 Shop All Cole-Parmer Products
Encompass_Preferred
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.
Catalog No. NC1476301 Supplier Cole-Parmer Supplier No. 5999401
Add to Cart
Add to Cart
These stainless steel (SS) testing sieves are serialized and come with a test sieve certificate of compliance. Equivalent to ASTM E11-09 compliance sieve.