Learn More
Cole-Parmer Cole-Parmer 3"-Diameter Sieve, Full Height Stainless Steel, No. 400 (38µ)
Catalog No. NC0265693 Shop All Cole-Parmer Products
Encompass_Preferred
Made of stainless steel; 3" diameter, full height sieve, 1" stacking height.
Catalog No. NC0265693 Supplier Cole-Parmer Supplier No. 5998730
Made of stainless steel; 3" diameter, full height sieve, 1" stacking height.