Learn More
Gilson Company Inc 3in (76mm) diameter Stainless Steel Sieve Cover
Catalog No. 50871325
Encompass_Preferred
3in (76mm) diameter Stainless Steel Sieve Cover for use with ASTM E11 3in Test Sieves or Pans prevents sample loss or contamination during the sieving process. Features: Fits all 3in Test Sieves or Pans, Prevents particle loss or contamination during sieving. Included Items: 3in All Stainless Sieve Cover.
Catalog No. 50-871-325 Supplier Gilson Company Inc Supplier No. V3SFXCV
3in (76mm) diameter Stainless Steel Sieve Cover for use with ASTM E11 3in Test Sieves or Pans prevents sample loss or contamination during the sieving process. Features: Fits all 3in Test Sieves or Pans, Prevents particle loss or contamination during sieving. Included Items: 3in All Stainless Sieve Cover.