Learn More
Gilson Company Inc V3SFXCR 3in (76mm) Stainless Steel Sieve Cover with Ring.
Catalog No. 50871324
Encompass_Preferred
V3SFXCR 3in (76mm) Stainless Steel Sieve Cover with Ring is for use with ASTM E11 3in Test Sieves or 3in Pans. The cover prevents sample loss or contamination during the sieving process. Built-in ring loop allows easy removal. Features: Fits all 3in test sieves or pans. Ring allows for easy removal of cover. 3in All Stainless Sieve Cover with Ring.
Catalog No. 50-871-324 Supplier Gilson Company Inc Supplier No. V3SFXCR
V3SFXCR 3in (76mm) Stainless Steel Sieve Cover with Ring is for use with ASTM E11 3in Test Sieves or 3in Pans. The cover prevents sample loss or contamination during the sieving process. Built-in ring loop allows easy removal. Features: Fits all 3in test sieves or pans. Ring allows for easy removal of cover. 3in All Stainless Sieve Cover with Ring.